siRNA Details
Gene Symbol: Sertad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108351.1 | NP_001101821.1 | 4 | 1148 | ||
XM_008769863.1 | XP_008768085.1 | 3 | 5039 | |||
XM_008769864.1 | XP_008768086.1 | 4 | 4509 | |||
GenBank mRNA | FQ217356.1 | - | 1 | 610 |
