siRNA Details
Gene Symbol: Fgfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199114.1 | NP_954545.1 | 2 | 307 | ||
GenBank mRNA | AJ536020.1 | - | 2 | 307 | ||
BC097261.1 | - | 2 | 263 | 7442454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
