siRNA Details
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100898.1 | NP_001094368.1 | 13 | 1540 | ||
XM_006250569.2 | XP_006250631.1 | 14 | 1735 | |||
XM_008769939.1 | XP_008768161.1 | 13 | 1445 | |||
GenBank mRNA | BC160928.1 | - | 11 | 1109 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001253877, NM_001253878, NM_001253879, NM_013827, NM_001253880, and others.
Gene Ontology Categories:
