siRNA Details
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100898.1 | NP_001094368.1 | 9 | 1055 | ||
XM_006250569.2 | XP_006250631.1 | 10 | 1250 | |||
XM_008769939.1 | XP_008768161.1 | 9 | 960 | |||
GenBank mRNA | BC093403.1 | - | 9 | 1015 | ||
BC160928.1 | - | 7 | 624 | |||
FQ227583.1 | - | 10 | 1252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
