siRNA Details
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108352.1 | NP_001101822.1 | 11 | 1432 | ||
XM_006250570.2 | XP_006250632.1 | 9 | 1168 | |||
XM_008769940.1 | XP_008768162.1 | 9 | 1145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
