siRNA Details
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004275.1 | NP_001004275.1 | 8 | 980 | ||
XM_006250667.2 | XP_006250729.1 | 7 | 890 | |||
XM_006250668.2 | XP_006250730.1 | 8 | 1026 | |||
GenBank mRNA | BC079384.1 | - | 8 | 980 | 7111425 |

More Information
Other Species Targeted

Gene Ontology Categories:
