siRNA Details
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004275.1 | NP_001004275.1 | 5 | 571 | ||
XM_006250667.2 | XP_006250729.1 | 4 | 481 | |||
XM_006250668.2 | XP_006250730.1 | 5 | 617 | |||
GenBank mRNA | BC079384.1 | - | 5 | 571 | 7111425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
