siRNA Details
Gene Symbol: LOC360919 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751340.3 | XP_003751388.1 | 4, 5 | 672 | ||
XM_006250759.2 | XP_006250821.1 | 4, 5 | 670 | |||
XM_006250760.2 | XP_006250822.1 | 4, 5 | 675 | |||
XM_006250761.2 | XP_006250823.1 | 4, 5 | 678 | |||
GenBank mRNA | BC088176.1 | - | 4, 5 | 507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
