siRNA Details
Gene Symbol: Pcdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004087.1 | NP_001004087.1 | 3 | 3526 | ||
XM_006251017.2 | XP_006251079.1 | 3 | 3628 | |||
GenBank mRNA | AY690615.1 | - | 3 | 3550 | ||
AY690616.1 | - | 3 | 3526 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001122758, AY861419, AB011255, AK039737, AY861420.
Gene Ontology Categories:
