siRNA Details
Gene Symbol: Pcdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004087.1 | NP_001004087.1 | 1 | 785 | ||
XM_006251017.2 | XP_006251079.1 | 1 | 863 | |||
XM_006251022.2 | XP_006251084.1 | 1 | 863 | |||
XM_006251023.2 | XP_006251085.1 | 1 | 863 | |||
XM_008770189.1 | XP_008768411.1 | 1 | 863 | |||
GenBank mRNA | AY690613.1 | - | 1 | 785 | ||
AY690614.1 | - | 1 | 785 | |||
AY690615.1 | - | 1 | 785 | |||
AY690616.1 | - | 1 | 785 |
