siRNA Details
Gene Symbol: Ablim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001514.2 | NP_001001514.1 | 15, 17 | 1687 | ||
XM_006251138.2 | XP_006251200.1 | 16, 18 | 2342 | |||
XM_008770329.1 | XP_008768551.1 | 15, 17 | 2242 | |||
XM_008770330.1 | XP_008768552.1 | 17, 19 | 2399 | |||
GenBank mRNA | AJ703893.2 | - | 15, 17 | 1687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
