siRNA Details
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004424.1 | NP_001004424.1 | 11 | 1423 | ||
GenBank mRNA | CB736453.1 | - | 4 | 288 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004424.1 | NP_001004424.1 | 11 | 1423 | ||
GenBank mRNA | CB736453.1 | - | 4 | 288 |