siRNA Details
Gene Symbol: Rnf185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024271.1 | NP_001019442.1 | 3, 4 | 357 | ||
XM_006251333.1 | XP_006251395.1 | 3, 4 | 325 | |||
XM_008770331.1 | XP_008768553.1 | 1, 2 | 169 | |||
GenBank mRNA | BC092655.1 | - | 3, 4 | 357 | 7320514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
