siRNA Details
Gene Symbol: Nipsnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100730.1 | NP_001094200.1 | 8 | 704 | ||
XM_006251337.2 | XP_006251399.1 | 8 | 1477 | |||
GenBank mRNA | BC089879.1 | - | 8 | 686 |

More Information
Other Species Targeted

Gene Ontology Categories:
