siRNA Details
Gene Symbol: Gas2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108365.1 | NP_001101835.1 | 6 | 1240 | ||
XM_006251340.2 | XP_006251402.1 | 5 | 1275 | |||
XM_008770332.1 | XP_008768554.1 | 4 | 1601 | |||
XM_008770333.1 | XP_008768555.1 | 3 | 1503 |
