siRNA Details
Gene Symbol: Zswim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271275.1 | NP_001258204.1 | 5 | 1012 | ||
XM_008770546.1 | XP_008768768.1 | 5 | 1024 | |||
XM_008770547.1 | XP_008768769.1 | 5 | 1024 | |||
XM_008770548.1 | XP_008768770.1 | 5 | 1023 | |||
XM_008770549.1 | XP_008768771.1 | 5 | 1024 | |||
XM_008770550.1 | XP_008768772.1 | 5 | 1024 | |||
XM_008770551.1 | XP_008768773.1 | 5 | 1024 |
Gene Symbol: LOC103689945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770640.1 | XP_008768862.1 | 4 | 547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
