siRNA Details
Gene Symbol: Ngly1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014136.1 | NP_001014158.1 | 9 | 1378 | ||
GenBank mRNA | BC083837.1 | - | 9 | 1378 | 7113173 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021504, BC028961, AK028248, AF250927, AK148931.
Gene Ontology Categories:
