siRNA Details
Gene Symbol: Mapk1ip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108373.1 | NP_001101843.1 | 3 | 768 | ||
XM_006251812.2 | XP_006251874.1 | 3 | 766 | |||
GenBank mRNA | BC167004.1 | - | 3 | 766 | 9090872 | |
FQ212003.1 | - | 1 | 571 |
