siRNA Details
Gene Symbol: RGD1310110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108374.1 | NP_001101844.1 | 4 | 1898 | ||
GenBank mRNA | FQ212344.1 | - | 4 | 1900 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1310110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108374.1 | NP_001101844.1 | 4 | 1898 | ||
GenBank mRNA | FQ212344.1 | - | 4 | 1900 |