siRNA Details
Gene Symbol: RGD1308430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108376.2 | NP_001101846.1 | 2 | 327 | ||
XM_006251977.2 | XP_006252039.1 | 2 | 247 | |||
XM_006251978.2 | XP_006252040.1 | 1 | 189 | |||
XM_008770680.1 | XP_008768902.1 | 2 | 594 | |||
GenBank mRNA | BC158656.1 | - | 2 | 327 | 8369089 | |
FQ219034.1 | - | 2 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
