siRNA Details
Gene Symbol: Parp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770809.1 | XP_008769031.1 | 32 | 5023 | ||
GenBank mRNA | BC086332.1 | - | 1 | 201 | ||
FQ235336.1 | - | 1 | 315 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Parp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770809.1 | XP_008769031.1 | 32 | 5023 | ||
GenBank mRNA | BC086332.1 | - | 1 | 201 | ||
FQ235336.1 | - | 1 | 315 |