siRNA Details
Gene Symbol: Parp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770809.1 | XP_008769031.1 | 33, 34 | 5273 | ||
GenBank mRNA | BC086332.1 | - | 2, 3 | 451 | ||
FQ235336.1 | - | 2, 3 | 565 |

More Information
Other Species Targeted

Gene Ontology Categories:
