siRNA Details
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108379.1 | NP_001101849.1 | 6, 7 | 1231 | ||
XM_006252074.1 | XP_006252136.1 | 6, 7 | 1037 | |||
XM_006252075.1 | XP_006252137.1 | 6, 7 | 1037 | |||
XM_008770758.1 | XP_008768980.1 | 6, 7 | 1034 | |||
GenBank mRNA | BC166461.1 | - | 6, 7 | 1008 | 9030107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
