siRNA Details
Gene Symbol: RGD1563302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162899.1 | NP_001156371.1 | 1 | 341 | ||
XM_006252122.2 | XP_006252184.1 | 1 | 395 | |||
GenBank mRNA | BF566484.1 | - | 1 | 267 | ||
BM422867.1 | - | 1 | 348 |
