siRNA Details
Gene Symbol: Ebpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108381.1 | NP_001101851.1 | 2 | 248 | ||
GenBank mRNA | FQ213140.1 | - | 2 | 254 |

More Information
Other Species Targeted

Gene Ontology Categories:
