siRNA Details
Gene Symbol: Rnaseh2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007007.1 | NP_001007008.1 | 11 | 879 | ||
XM_008770762.1 | XP_008768984.1 | 11 | 2952 | |||
XM_008770764.1 | XP_008768986.1 | 10 | 2873 | |||
GenBank mRNA | BC083791.1 | - | 11 | 879 | 7106440 | |
FQ220108.1 | - | 11 | 906 |
