siRNA Details
Gene Symbol: RGD1308117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134571.2 | NP_001128043.2 | 2 | 195 | ||
XM_006252278.2 | XP_006252340.1 | 2 | 2284 | |||
XM_008770829.1 | XP_008769051.1 | 1 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
