siRNA Details
Gene Symbol: Nudt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100732.1 | NP_001094202.1 | 3 | 544 | ||
GenBank mRNA | BC082050.1 | - | 477 | |||
BC098710.1 | - | 3 | 546 | |||
BC158874.1 | - | 3 | 544 | 7933460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
