siRNA Details
Gene Symbol: Dmtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108385.1 | NP_001101855.1 | 10 | 1160 | ||
XM_006252279.2 | XP_006252341.1 | 9 | 1295 | |||
XM_006252280.2 | XP_006252342.1 | 9 | 1293 | |||
XM_006252281.1 | XP_006252343.1 | 7 | 648 | |||
XM_008770830.1 | XP_008769052.1 | 10 | 1158 | |||
XM_008770831.1 | XP_008769053.1 | 10 | 1815 | |||
GenBank mRNA | BC166453.1 | - | 9 | 1169 | 8374835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
