siRNA Details
Gene Symbol: Xpo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108386.2 | NP_001101856.2 | 9 | 1045 | ||
XM_006252282.2 | XP_006252344.1 | 9 | 2558 | |||
XM_008770832.1 | XP_008769054.1 | 9 | 2601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
