siRNA Details
Gene Symbol: Xpo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108386.2 | NP_001101856.2 | 19 | 2153 | ||
XM_006252282.2 | XP_006252344.1 | 19 | 3666 | |||
XM_008770832.1 | XP_008769054.1 | 19 | 3709 |

More Information
Other Species Targeted

Gene Ontology Categories:
