siRNA Details
Gene Symbol: Slc25a30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013187.1 | NP_001013205.1 | 2, 3 | 191 | ||
XM_008770873.1 | XP_008769095.1 | 2, 3 | 191 | |||
GenBank mRNA | AY724532.1 | - | 2, 3 | 163 | ||
BC087106.1 | - | 2, 3 | 191 | 7193157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
