siRNA Details
Gene Symbol: Ndfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108390.2 | NP_001101860.2 | 7 | 1039 | ||
XM_006252429.1 | XP_006252491.1 | 8 | 984 | |||
GenBank mRNA | DY565248.1 | - | 6 | 601 | ||
FM044778.1 | - | 2 | 88 |
