siRNA Details
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034937.2 | NP_001030109.2 | 8 | 980 | ||
XM_006252509.2 | XP_006252571.1 | 7 | 1006 | |||
GenBank mRNA | BC105842.1 | - | 3 | 363 | 7455960 |

More Information
Other Species Targeted

Gene Ontology Categories:
