siRNA Details
Gene Symbol: RGD1309676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014140.1 | NP_001014162.1 | 3 | 283 | ||
XM_006252791.2 | XP_006252853.1 | 3 | 1085 | |||
GenBank mRNA | BC079275.1 | - | 3 | 283 | 7134042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
