siRNA Details
Gene Symbol: Eps15l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029921.1 | NP_001025092.1 | 14, 15 | 1443 | ||
XM_006252802.2 | XP_006252864.1 | 14, 15 | 1594 | |||
XM_006252804.2 | XP_006252866.1 | 14, 15 | 1592 | |||
XM_008771118.1 | XP_008769340.1 | 14, 15 | 1581 | |||
XM_008771119.1 | XP_008769341.1 | 14, 15 | 1581 | |||
XM_008771120.1 | XP_008769342.1 | 14, 15 | 1580 | |||
XM_008771121.1 | XP_008769343.1 | 14, 15 | 1580 | |||
XM_008771122.1 | XP_008769344.1 | 14, 15 | 1580 | |||
XM_008771123.1 | XP_008769345.1 | 14, 15 | 1579 | |||
GenBank mRNA | BC098004.1 | - | 14, 15 | 1443 | 7453598 |
