siRNA Details
Gene Symbol: Tmem192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014141.1 | NP_001014163.1 | 3, 4 | 634 | ||
GenBank mRNA | BC086402.1 | - | 3, 4 | 634 | 7304126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
