siRNA Details
Gene Symbol: Tmem192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014141.1 | NP_001014163.1 | 3, 4 | 627 | ||
GenBank mRNA | BC086402.1 | - | 3, 4 | 627 | 7304126 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014141.1 | NP_001014163.1 | 3, 4 | 627 | ||
GenBank mRNA | BC086402.1 | - | 3, 4 | 627 | 7304126 |