siRNA Details
Gene Symbol: Primpol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253167.2 | XP_006253229.1 | 7 | 771 | ||
XM_341433.6 | XP_341434.3 | 7 | 768 | |||
GenBank mRNA | FQ230438.1 | - | 4 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
