siRNA Details
Gene Symbol: Bag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025130.1 | NP_001020301.1 | 4 | 1354 | ||
GenBank mRNA | BC083608.1 | - | 4 | 1354 | 7133487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
