siRNA Details
Gene Symbol: Adam32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170582.1 | NP_001164053.1 | 16 | 1675 | ||
XM_008771361.1 | XP_008769583.1 | 16 | 1785 | |||
XM_008771362.1 | XP_008769584.1 | 16 | 2090 | |||
GenBank mRNA | AJ131563.1 | - | 11 | 1223 |
