siRNA Details
Gene Symbol: Golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007731.1 | NP_001007732.1 | 2 | 359 | ||
XM_006253320.2 | XP_006253382.1 | 2 | 407 | |||
GenBank mRNA | BC078959.1 | - | 2 | 359 | 7110849 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_020585, NM_001042484, BC016894, AK153213, AK150462, and others.
Gene Ontology Categories:
