siRNA Details
Gene Symbol: Alg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108401.1 | NP_001101871.1 | 3 | 977 | ||
XM_006253354.2 | XP_006253416.1 | 3 | 1249 | |||
XM_006253355.2 | XP_006253417.1 | 3 | 951 | |||
XM_006253356.2 | XP_006253418.1 | 3 | 1574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
