siRNA Details
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253453.2 | XP_006253515.1 | 5 | 1166 | ||
XM_006253454.2 | XP_006253516.1 | 5 | 1507 | |||
XM_006253458.2 | XP_006253520.1 | 5 | 1606 | |||
XM_006253459.2 | XP_006253521.1 | 3 | 1306 | |||
XM_008771402.1 | XP_008769624.1 | 5 | 5314 | |||
GenBank mRNA | BC098779.1 | - | 1 | 29 |
