siRNA Details
Gene Symbol: Asb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044247.1 | NP_001037712.1 | 3, 4 | 460 | ||
XM_006253100.2 | XP_006253162.1 | 3, 4 | 1890 | |||
XM_008771246.1 | XP_008769468.1 | 3, 4 | 893 | |||
XM_008771247.1 | XP_008769469.1 | 3, 4 | 488 | |||
XM_008771248.1 | XP_008769470.1 | 3, 4 | 1733 | |||
GenBank mRNA | BC098935.1 | - | 3, 4 | 460 | 7445963 | |
FQ214804.1 | - | 3, 4 | 410 | |||
FQ215658.1 | - | 3, 4 | 332 |
