siRNA Details
Gene Symbol: LOC103693963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771279.1 | XP_008769501.1 | 1 | 210 |
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772487.1 | XP_008770709.1 | 1 | 157 |
