siRNA Details
Gene Symbol: Usp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001516.2 | NP_001001516.1 | 15 | 2113 | ||
XM_006243793.2 | XP_006243855.1 | 16 | 2431 | |||
XM_006243794.2 | XP_006243856.1 | 16 | 2428 | |||
XM_006243795.2 | XP_006243857.1 | 16 | 2425 | |||
XM_006243796.2 | XP_006243858.1 | 16 | 2407 | |||
XM_006243797.2 | XP_006243859.1 | 16 | 2430 | |||
XM_006243798.2 | XP_006243860.1 | 16 | 2427 | |||
XM_006243799.2 | XP_006243861.1 | 16 | 2424 | |||
GenBank mRNA | AY605065.1 | - | 15 | 2113 | ||
BC168243.1 | - | 15 | 2439 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001168371, NM_027804, NM_001168373, NM_001168372, NM_145407, and others.
Gene Ontology Categories:
