siRNA Details
Gene Symbol: Usp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001516.2 | NP_001001516.1 | 6, 7 | 937 | ||
XM_006243793.2 | XP_006243855.1 | 7, 8 | 1249 | |||
XM_006243794.2 | XP_006243856.1 | 7, 8 | 1246 | |||
XM_006243795.2 | XP_006243857.1 | 7, 8 | 1249 | |||
XM_006243796.2 | XP_006243858.1 | 7, 8 | 1225 | |||
XM_006243797.2 | XP_006243859.1 | 7, 8 | 1248 | |||
XM_006243798.2 | XP_006243860.1 | 7, 8 | 1245 | |||
XM_006243799.2 | XP_006243861.1 | 7, 8 | 1248 | |||
GenBank mRNA | AY605065.1 | - | 6, 7 | 937 | ||
BC168243.1 | - | 6, 7 | 1257 |
