siRNA Details
Gene Symbol: Habp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079940.1 | NP_001073409.1 | 6 | 957 | ||
XM_006253494.2 | XP_006253556.1 | 5 | 853 | |||
GenBank mRNA | BC129111.1 | - | 6 | 957 | 7111244 |

More Information
Other Species Targeted

Gene Ontology Categories:
