siRNA Details
Gene Symbol: Fbxw17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082409.1 | NP_001075878.1 | 4 | 359 | ||
XM_008771539.1 | XP_008769761.1 | 4 | 404 | |||
XM_008771540.1 | XP_008769762.1 | 4 | 404 | |||
XM_008771541.1 | XP_008769763.1 | 4 | 404 | |||
XM_008771542.1 | XP_008769764.1 | 4 | 1366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
